Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Universal Pipette Tips, Bulk Packed
SCI_ED

Catalog No. S317183A Shop All Fischer Technical Company Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
5000 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S317183A 5000 μL
S012387 10 μL
S012388 200 μL
S06825 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S317183A Supplier Fischer Technical Company™ Supplier No. D893PK 500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Universal Pipette Tips provide volumetric accuracy for your pipetting needs.

  • Ideal molded quality
  • Good fit for accurate dispensing
  • Bulk-packed
  • Non-sterile
  • Suitable for most leading brands

Specifications

Volume (Metric) 5000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Bulk, Bagged
Tip Style Standard
Quantity 500 Tips/Pk.
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Plastic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.