Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Multi Function 3D Rotators

Catalog No. 88861048 Shop All Fisherbrand Products
Change view
Click to view available options
Plug Type:
EU, UK, CN Plug
US Plug
Certifications/Compliance:
CE
cCSAus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Certifications/Compliance
88-861-048 EU, UK, CN Plug CE
88-861-047 US Plug cCSAus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88-861-048
Only null left
Add to Cart
Add to Cart

Includes

Dimpled rubber mat and six tube-securing bands

For continuous operation, timed operation or different combinations to ensure optimal mixing conditions for any application

  • Rotation speed: 5 to 100 rpm for long wave motion
  • Reciprocal rotation: 0°to 360° turning angle for long and medium way motion
  • Vibration: 0° to 5° turning angle, programmable in bursts of 0 to 5 seconds, for short wave motion
  • Fits neatly into small workspaces
  • Operating temperature range of 5 to 40°C allows for use in refrigerators or non-CO2 incubators
  • Continuous or timed operation with automatic switch-off
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Audible and visual alarm: In timed mode, when the time reaches zero, alarm will sound 5 times, then the time window shows“End“ and flickers all the time. Alarm can be stopped by touch any key.

Certifications

CE

Warranty and Services

2 years

TRUSTED_SUSTAINABILITY

Specifications

Orbit 23 mm
Rotating Angle 0 to 360°
Load Bearing Capacity (Metric) 1 kg
Speed Range 5 to 100 rpm
Voltage 100/240 V
Temperature Range (Metric) 5°C to 40°C
Load Bearing Capacity (English) 2.2 lb.
Certifications/Compliance CE
Plug Type EU, UK, CN Plug
Length (Metric) Platform 306 mm
Width (Metric) Platform 266 mm
For Use With (Application) Immune precipitations, Affinity matrix , Trypsinisation, Gel straining and distaining, Antibody staining, Washes, Hybridizations, Southern blots, Western blots
Timer Range
  • Orbital/360° vertical/reciprocal mode - 0 to 250 sec.
  • Vibration mode - 0- 5 sec (Vibration) with 0- 125 cycles
Warranty and Service Offering 2 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.