Learn More
Chemglass Life Sciences Multi-Function Liquid Transfer Cap for 3 L wide-mouth and 5 L high-efficiency Erlenmeyer flasks
SDP
Catalog No. 502733448
Encompass
The multi-function liquid transfer cap is designed for aseptic liquid transfer and sampling with 3 L wide-mouth and 5 L high-efficiency Erlenmeyer flasks. It supports direct placement in an incubator after liquid transfer, provides a large air flux, and includes a sampling nozzle and inlet filtration to reduce contamination and minimize liquid retention in tubing.
  • Can be placed in an incubator after liquid transfer
  • Provides large air flux
  • Sampling nozzle with one-way valve prevents liquid backflow during aseptic sampling
  • Liquid inlet tubing features a PTFE needle filter
  • Compatible with 3 L wide-mouth and 5 L high-efficiency Erlenmeyer flasks
  • Pack quantity: CS4 (manufacturer designation)
Catalog No. 50-273-3448 Supplier Chemglass Life Sciences Supplier No. CGN2165787
Add to Cart
Add to Cart
The multi-function liquid transfer cap is designed for aseptic liquid transfer and sampling with 3 L wide-mouth and 5 L high-efficiency Erlenmeyer flasks. It supports direct placement in an incubator after liquid transfer, provides a large air flux, and includes a sampling nozzle and inlet filtration to reduce contamination and minimize liquid retention in tubing.
  • Can be placed in an incubator after liquid transfer
  • Provides large air flux
  • Sampling nozzle with one-way valve prevents liquid backflow during aseptic sampling
  • Liquid inlet tubing features a PTFE needle filter
  • Compatible with 3 L wide-mouth and 5 L high-efficiency Erlenmeyer flasks
  • Pack quantity: CS4 (manufacturer designation)