Learn More
Chemglass Life Sciences Multi-Function Liquid Transfer Cap for Nest 3L High-Efficiency Erlenmeyer Flasks
SDP
Catalog No. 502733449
Encompass
The Multi-Function Liquid Transfer Cap enables aseptic liquid transfer and convenient sampling for 3 L high-efficiency Erlenmeyer flasks. It supports direct placement in an incubator after transfer, provides high air flux, and prevents backflow during sampling with an integrated one-way valve and PTFE-filtered inlet tubing.
  • Allows direct placement in incubator after transfer.
  • Provides high air flux to support culture aeration.
  • Sampling nozzle with one-way valve prevents backflow during sampling.
  • Inlet tubing includes PTFE needle filter to reduce residual liquid in tubing.
  • Packaged in case quantity of four.
Catalog No. 50-273-3449 Supplier Chemglass Life Sciences Supplier No. CGN2165786
Add to Cart
Add to Cart
The Multi-Function Liquid Transfer Cap enables aseptic liquid transfer and convenient sampling for 3 L high-efficiency Erlenmeyer flasks. It supports direct placement in an incubator after transfer, provides high air flux, and prevents backflow during sampling with an integrated one-way valve and PTFE-filtered inlet tubing.
  • Allows direct placement in incubator after transfer.
  • Provides high air flux to support culture aeration.
  • Sampling nozzle with one-way valve prevents backflow during sampling.
  • Inlet tubing includes PTFE needle filter to reduce residual liquid in tubing.
  • Packaged in case quantity of four.