Learn More
Chemglass Life Sciences Multi-Function Liquid Transfer Cap for 2 L High Efficiency Erlenmeyer Flasks
SDP
Catalog No. 502733450
Encompass
Multi-function liquid transfer cap designed for 2 L shake flasks. It enables aseptic liquid transfer and supports post-transfer incubation, providing high air flux and features to minimize liquid retention.
  • Can be placed in an incubator after liquid transfer.
  • Provides high air flux during transfer.
  • Sampling nozzle includes a one-way valve to prevent backflow.
  • Liquid inlet tubing includes a PTFE needle filter to reduce liquid retention.
  • Compatible with 2 L flasks; sold in cases of four.
Catalog No. 50-273-3450 Supplier Chemglass Life Sciences Supplier No. CGN2165785
Add to Cart
Add to Cart
Multi-function liquid transfer cap designed for 2 L shake flasks. It enables aseptic liquid transfer and supports post-transfer incubation, providing high air flux and features to minimize liquid retention.
  • Can be placed in an incubator after liquid transfer.
  • Provides high air flux during transfer.
  • Sampling nozzle includes a one-way valve to prevent backflow.
  • Liquid inlet tubing includes a PTFE needle filter to reduce liquid retention.
  • Compatible with 2 L flasks; sold in cases of four.