Learn More
Electron Microscopy Sciences Multi Holder for 29 pin Stubs, Hitachi M4
SDP
Catalog No. 501901518
Encompass
This 90 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place. It accommodates up to twenty nine standard 12.7 mm (1/2") pin stubs
Type: Hitachi M4
Catalog No. 50-190-1518 Supplier Electron Microscopy Sciences Supplier No. 7595229
This 90 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place. It accommodates up to twenty nine standard 12.7 mm (1/2") pin stubs
Type: Hitachi M4