Learn More
Electron Microscopy Sciences Multi Holder for 8 Pin Stubs, 3.2 mm x 8mm Pin
SDP
Catalog No. 501901514
Encompass
This 50 mm in diameter multi pin holder is designed to save time.It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (1/2") pin stubs.
Type: 3.2 mm
Catalog No. 50-190-1514 Supplier Electron Microscopy Sciences Supplier No. 7595208
This 50 mm in diameter multi pin holder is designed to save time.It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (1/2") pin stubs.
Type: 3.2 mm