Learn More
Analytical Sales & Services Multi-Inlet Waste Manifold with 4 Standard Tubing Ports, 6 Ports for Barbed Adapter
Catalog No. 5000806188
Encompass
Canary-Safe Multi-Inlet Waste Manifold with 4 Standard Tubing Ports for 1/16" or 1/8" OD Tubing, 6 Ports for Barbed Adapter and 1 Large Outlet. Fits Benches up to 3" Thick. Includes Fittings for 1/8" tubing and Barbed Adapters. Fittings for 1/16" tubing sold separately, see part # 59249.
Catalog No. 50-008-06188 Supplier Analytical Sales & Services Supplier No. MW0406
Canary-Safe Multi-Inlet Waste Manifold with 4 Standard Tubing Ports for 1/16" or 1/8" OD Tubing, 6 Ports for Barbed Adapter and 1 Large Outlet. Fits Benches up to 3" Thick. Includes Fittings for 1/8" tubing and Barbed Adapters. Fittings for 1/16" tubing sold separately, see part # 59249.