Learn More
Analytical Sales & Services MULTI-INLET WST MANIFOLD 5PRT
Catalog No. NC2200067
Encompass
multi-inlet waste manifold with 5 standard tubing ports, 2 ports for 3/16" od tubing, 3 ports for barbed adapters
This item is not returnable. View return policy
Catalog No. NC2200067 Supplier Analytical Sales & Services Supplier No. MW0523
multi-inlet waste manifold with 5 standard tubing ports, 2 ports for 3/16" od tubing, 3 ports for barbed adapters