Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multi-Ion Standard, Ricca Chemical
SDP

Catalog No. MSTD2004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSTD200-4 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSTD200-4 Supplier Ricca Chemical Company Supplier No. MSTD2004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

100 ppm each Ca2+, Mg2+, Na+, NH4+, Li+, K+ for Ion Chromatography

Specifications

Chemical Name or Material Multi-Ion Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5, 7447-41-8, 10035-04-8, 12125-02-9
Concentration or Composition Notes 100ppm each Ca
CAS Min % 98.0, 0.08, 0.06, 0.04, 0.03, 0.02
CAS Max % 100.0, 0.09, 0.06, 0.04, 0.03, 0.03
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Ion Chromatography Standard
Quantity 120 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.