Learn More
Apexbio Technology LLC Multi-kinase inhibitor 1 778274-97-8 1mg
SDP
Catalog No. 5000096135
Encompass_Preferred
Multi-kinase inhibitor 1 (CAS 778274-97-8) is a small molecule designed to concurrently inhibit multiple receptor tyrosine kinases including VEGFR PDGFR FGFR and c-Kit By targeting these signaling pathways the compound interferes with processes fundamental to tumor cell proliferation angiogenesis and metastasis Multi-kinase inhibitor 1 is utilized in biomedical research to explore the roles of these kinases in cancer biology model multi-targeted therapeutic strategies and assess the impact of broad-spectrum kinase inhibition on tumor growth and progression
Catalog No. 50-000-96135 Supplier Apexbio Technology LLC Supplier No. A99561
Multi-kinase inhibitor 1 (CAS 778274-97-8) is a small molecule designed to concurrently inhibit multiple receptor tyrosine kinases including VEGFR PDGFR FGFR and c-Kit By targeting these signaling pathways the compound interferes with processes fundamental to tumor cell proliferation angiogenesis and metastasis Multi-kinase inhibitor 1 is utilized in biomedical research to explore the roles of these kinases in cancer biology model multi-targeted therapeutic strategies and assess the impact of broad-spectrum kinase inhibition on tumor growth and progression