Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Multi-kinase inhibitor 1 | 778274-97-8 | 99.1% | 418.37 g·mol⁻¹ | C20H17F3N4O3 | 1 ML
SDP

Catalog No. 5000245978
Encompass_Preferred

Multi-kinase inhibitor 1 is a small-molecule multi-kinase inhibitor provided as a ready-to-use 10 mM solution in DMSO for laboratory research. It has molecular formula C20H17F3N4O3, a molecular weight of 418.37 g·mol⁻¹, and CAS number 778274-97-8. Intended for research use only.

  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.05%).
  • Molecular weight 418.37 g·mol⁻¹.
  • CAS 778274-97-8.
  • Supplied as 1 mL vial for single-use experiments.

Catalog No. 50-002-45978 Supplier Medchemexpress LLC Supplier No. HY10303210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Multi-kinase inhibitor 1 is a small-molecule multi-kinase inhibitor provided as a ready-to-use 10 mM solution in DMSO for laboratory research. It has molecular formula C20H17F3N4O3, a molecular weight of 418.37 g·mol⁻¹, and CAS number 778274-97-8. Intended for research use only.

  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.05%).
  • Molecular weight 418.37 g·mol⁻¹.
  • CAS 778274-97-8.
  • Supplied as 1 mL vial for single-use experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.