Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Multi-Leu peptide triacetate | 98.6% | 1231.53 | 100 MG
SDP

Catalog No. 5000423216
Encompass_Preferred

Multi-Leu peptide triacetate (ML-peptide) is a potent inhibitor of PACE4 with a Ki of 22 nM. It competitively binds to the active site of PACE4 by simulating the substrate sequence, thereby inhibiting its catalytic activity. This peptide can be used to study the specific mechanism of PACE4 in the development of prostate cancer.

  • Potent inhibitor of PACE4 (Ki=22 nM)
  • Competitively binds to active site of PACE4
  • Inhibits catalytic activity of PACE4
  • Useful for studying PACE4 mechanism in prostate cancer development
  • Molecular weight of 1231.53
  • Purity of 98.6%

Catalog No. 50-004-23216 Supplier Medchemexpress LLC Supplier No. HYP10422A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Multi-Leu peptide triacetate (ML-peptide) is a potent inhibitor of PACE4 with a Ki of 22 nM. It competitively binds to the active site of PACE4 by simulating the substrate sequence, thereby inhibiting its catalytic activity. This peptide can be used to study the specific mechanism of PACE4 in the development of prostate cancer.

  • Potent inhibitor of PACE4 (Ki=22 nM)
  • Competitively binds to active site of PACE4
  • Inhibits catalytic activity of PACE4
  • Useful for studying PACE4 mechanism in prostate cancer development
  • Molecular weight of 1231.53
  • Purity of 98.6%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.