Learn More
RNA Medical MULTI-LVL 3LVLX10X2.5ML 30/PK
SDP
Catalog No. NC9436558
Encompass
Control; Multi-Level (30 Amp x 2.5mL,10 Amp each level); Blood Gas Electrolyte CO-Oximeter (Bayer and AVOX Systems) Control); CC 527 has a shelf life of 36 months from the date of manufacture when stored at 2deg. to 8deg.C
Catalog No. NC9436558 Supplier RNA Medical Supplier No. CC 527
Control; Multi-Level (30 Amp x 2.5mL,10 Amp each level); Blood Gas Electrolyte CO-Oximeter (Bayer and AVOX Systems) Control); CC 527 has a shelf life of 36 months from the date of manufacture when stored at 2deg. to 8deg.C