Learn More
Electron Microscopy Sciences Multi-Microplate Genie, Swiss Plug, 230 V
SDP
Catalog No. 5033708
Encompass_Preferred
The Microplate mixer has high speed action for increased mixing of up to 4 microplates (8 with optional accessory tray). Heat will not be transferred to your microplates. The fan (located in the bottom of the unit) directs air downward and out of the unit. As a result, room air is constantly drawn into the unit underneath the tray that holds the microplates, insuring that samples will not be heated.
Features aggressive mixing speed from 750-3200 RPM for complete and uniform mixing in any microplate format.
Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity.
Speed and orbit combine to offer true vortexing action in each well of a microplate.
Accepts most microplate types
Built-in timer operates from 1 - 60 minutes or continuous for hands-free use.
May be used in cold rooms and incubators for temperature sensitive applications.
Catalog No. 50-337-08 Supplier Electron Microscopy Sciences Supplier No. 61042SW
The Microplate mixer has high speed action for increased mixing of up to 4 microplates (8 with optional accessory tray). Heat will not be transferred to your microplates. The fan (located in the bottom of the unit) directs air downward and out of the unit. As a result, room air is constantly drawn into the unit underneath the tray that holds the microplates, insuring that samples will not be heated.
Features aggressive mixing speed from 750-3200 RPM for complete and uniform mixing in any microplate format.
Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity.
Speed and orbit combine to offer true vortexing action in each well of a microplate.
Accepts most microplate types
Built-in timer operates from 1 - 60 minutes or continuous for hands-free use.
May be used in cold rooms and incubators for temperature sensitive applications.