Learn More
Research Products International Corp Multi Miniflex Tips, Extra Short, Round Tip, 0.1-10 ul Capacity, 200 per Case
SDP
Catalog No. 50212375
Encompass
Ultra-thin microcapillary pipet tips are ideal for nucleic acid sequencing and protein separation. Extra short MiniFlex Tips offer precise volume control and minimize sampling errors. Features a 20 ml graduation for volume confirmation and a non-wettable surface to ensure precision in ultra micro ranges. The universal sealing ring allows use with all major ultra-micro pipettors such as Pipetman P2 and P10, Eppendorf, Oxford Benchmate and Socorex. They are constructed of nucleated polypropylene and are autoclavable to 120°C.
Catalog No. 50-212-375 Supplier Research Products International Corp Supplier No. 148606
Ultra-thin microcapillary pipet tips are ideal for nucleic acid sequencing and protein separation. Extra short MiniFlex Tips offer precise volume control and minimize sampling errors. Features a 20 ml graduation for volume confirmation and a non-wettable surface to ensure precision in ultra micro ranges. The universal sealing ring allows use with all major ultra-micro pipettors such as Pipetman P2 and P10, Eppendorf, Oxford Benchmate and Socorex. They are constructed of nucleated polypropylene and are autoclavable to 120°C.