Learn More
Molecular Devices Molecular Devices Microplate Reader, 2 Read Modes, 6-384 Well
Catalog No. 5000014393
Encompass_Preferred
The Molecular Devices SpectraMax Mini Multi-Mode Microplate Reader is a compact, configurable instrument designed for routine absorbance, fluorescence, and luminescence assays. It is offered in two-mode configurations and can be upgraded in the field to add a third detection mode, providing a cost-effective and flexible platform for low- to mid-throughput laboratories.
  • Two read modes with user-upgradeable option to add a third
  • Supports 6-384 well plate formats for flexible throughput
  • Software compatible for streamlined data acquisition and analysis
  • Compact footprint for benchtop use
  • Suitable for absorbance, fluorescence, and luminescence assays
Catalog No. 50-000-14393 Supplier Molecular Devices Supplier No. SMAXMINIAL
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices SpectraMax Mini Multi-Mode Microplate Reader is a compact, configurable instrument designed for routine absorbance, fluorescence, and luminescence assays. It is offered in two-mode configurations and can be upgraded in the field to add a third detection mode, providing a cost-effective and flexible platform for low- to mid-throughput laboratories.
  • Two read modes with user-upgradeable option to add a third
  • Supports 6-384 well plate formats for flexible throughput
  • Software compatible for streamlined data acquisition and analysis
  • Compact footprint for benchtop use
  • Suitable for absorbance, fluorescence, and luminescence assays