Learn More
Molecular Devices Molecular Devices Replacement Lamp Pack, Microplate Reader, 2-Lamp, 5 V
Catalog No. 5000014617
Encompass_Preferred
Replacement lamp pack for use with Molecular Devices Emax and Vmax microplate readers. Contains two 5 V lamps to restore the reader's illumination for optical assays.
  • Restores instrument light output for absorbance and fluorescence measurements.
  • Includes two replacement lamps per package for complete lamp replacement.
  • 5 V lamp specification designed for microplate reader lamp assemblies.
  • Field-replaceable bulbs simplify routine maintenance.
Catalog No. 50-000-14617 Supplier Molecular Devices Supplier No. 03101000R
Replacement lamp pack for use with Molecular Devices Emax and Vmax microplate readers. Contains two 5 V lamps to restore the reader's illumination for optical assays.
  • Restores instrument light output for absorbance and fluorescence measurements.
  • Includes two replacement lamps per package for complete lamp replacement.
  • 5 V lamp specification designed for microplate reader lamp assemblies.
  • Field-replaceable bulbs simplify routine maintenance.