Learn More
Amsbio LLC Multi-normal human tissuesarra
Catalog No. 502564469
Encompass
Multi-normal human tissuesarray representingFDAguideline forantibody cross-reactivity testing32 types of human organs99 cores each type taken from 3 different individuals(1.5mm) replacing FDA999w
Catalog No. 50-256-4469 Supplier Amsbio LLC Supplier No. FDA999W3
Multi-normal human tissuesarray representingFDAguideline forantibody cross-reactivity testing32 types of human organs99 cores each type taken from 3 different individuals(1.5mm) replacing FDA999w