Learn More
Electron Microscopy Sciences Multi Holder for 29 pin stubs, 3.2 mm x 8mm Pin
SDP
Catalog No. 501901519
Encompass
This 90 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place. It accommodates up to twenty nine standard 12.7 mm (1/2") pin stubs
Type: 3.2 mm x 8mm Pin
Catalog No. 50-190-1519 Supplier Electron Microscopy Sciences Supplier No. 7595229H
This 90 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place. It accommodates up to twenty nine standard 12.7 mm (1/2") pin stubs
Type: 3.2 mm x 8mm Pin