Learn More
Electron Microscopy Sciences Multi Pin Holder Hitachi, M4 for 49 stubs
SDP
Catalog No. 501901644
Encompass
This 117 mm in diameter multi pin holder is designed to save time It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place. It accommodates up to forty nine standard 12.7 mm (1/2") pin stubs.
Catalog No. 50-190-1644 Supplier Electron Microscopy Sciences Supplier No. 7595249H
This 117 mm in diameter multi pin holder is designed to save time It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place. It accommodates up to forty nine standard 12.7 mm (1/2") pin stubs.