Learn More
MSC Dow Corning 1891961 3 oz Tube Silver RTV Silicone Joint Sealant -76 to -29.2°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 501149223
Encompass
Dow Corning 1891961 3 oz Tube Silver RTV Silicone Joint Sealant -76 to -29.2°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 50-114-9223 Supplier MSC Supplier No. 00231704
Dow Corning 1891961 3 oz Tube Silver RTV Silicone Joint Sealant -76 to -29.2°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732