Learn More
Dino Lite RK-06FA Multi-purpose Tabletop Stand with Fine Adjustment
Catalog No. NC1757756
Encompass_Preferred
Dino-Lite RK-06FA is a multi-purpose tabletop stand that provides flexibility and robustness, yet provides fine adjustment to ease focusing tasks. Capable of Up & Down, Lateral Movement. Includes RK-10-EX arm for additional positional capabilities. For users that require additional working distance, the optional RK-06-PL extension pole is sold separately.
Catalog No. NC1757756 Supplier Dino Lite Supplier No. RK06FA
Dino-Lite RK-06FA is a multi-purpose tabletop stand that provides flexibility and robustness, yet provides fine adjustment to ease focusing tasks. Capable of Up & Down, Lateral Movement. Includes RK-10-EX arm for additional positional capabilities. For users that require additional working distance, the optional RK-06-PL extension pole is sold separately.