Learn More
A2Z OZONE INC A2Z Ozone - MP-1000 Ozone Generator - Corona Discharge Water Ozonator, 1000 mg/hour, Single-Run, Built-In Compressor, Optional Oxygen Hookup
SDP
Catalog No. NC0921616
Encompass_Preferred
The MP-1000 Ozone Generator paired with an oxygen generator gives up to 1000 mg/hour of ozone. The oxygen generator allows for a more pure form of ozone. Use a regulator for 1-3 L/minute oxygen gas flow. Without an oxygen generator, expect about 1/5 of the ozone. Special features include 15-minute to 4-hour adjustable timer. The MP-1000 Ozone Generator comes with a stainless steel handle for portability. It also comes ready to use, with 8-feet of flexible polyurethane tubing, 8-feet of ozone resistant PTFE tubing, two 2" oblong diffuser stones, spare fuse, and a replacement fan filter. Dimensions: 10.7x6x9 inches, 8.5 pounds. Energy consumption is 45 Watts. Ozone concentration is 0-14 gr/m3. NOTE: This unit does not have a continuous setting.
Catalog No. NC0921616 Supplier A2Z OZONE INC Supplier No. MP1000
The MP-1000 Ozone Generator paired with an oxygen generator gives up to 1000 mg/hour of ozone. The oxygen generator allows for a more pure form of ozone. Use a regulator for 1-3 L/minute oxygen gas flow. Without an oxygen generator, expect about 1/5 of the ozone. Special features include 15-minute to 4-hour adjustable timer. The MP-1000 Ozone Generator comes with a stainless steel handle for portability. It also comes ready to use, with 8-feet of flexible polyurethane tubing, 8-feet of ozone resistant PTFE tubing, two 2" oblong diffuser stones, spare fuse, and a replacement fan filter. Dimensions: 10.7x6x9 inches, 8.5 pounds. Energy consumption is 45 Watts. Ozone concentration is 0-14 gr/m3. NOTE: This unit does not have a continuous setting.