Learn More
Waters Corp Waters Rack Adapter, Multi-Purpose, Flexible Sample Collection
Catalog No. 50793101 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Multi-Purpose Rack Adapter for the Waters Fraction Collector III replaces the standard tube rack, offering flexible sample collection by accommodating various racks.
  • Replaces the standard tube rack.
  • Accepts a variety of racks.
  • Accommodates one Sample Manager rack.
  • Holds up to two Fraction Collector racks.
  • Compatible with up to eight test tube holders.
  • Test tube holders can hold eight or twelve tubes.
  • Can be used with a combination of one Fraction Collector rack and up to four test tube holders.
Catalog No. 50-793-101 Supplier Waters Corp Supplier No. 725000120
The Multi-Purpose Rack Adapter for the Waters Fraction Collector III replaces the standard tube rack, offering flexible sample collection by accommodating various racks.
  • Replaces the standard tube rack.
  • Accepts a variety of racks.
  • Accommodates one Sample Manager rack.
  • Holds up to two Fraction Collector racks.
  • Compatible with up to eight test tube holders.
  • Test tube holders can hold eight or twelve tubes.
  • Can be used with a combination of one Fraction Collector rack and up to four test tube holders.