Learn More
Waters Corp Waters Corporation Rack, Multi-Purpose, Preparative Funnel Adapter
Catalog No. 50793100 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Multi-Purpose Rack is a versatile component designed for laboratory use, functioning as a preparative funnel adapter for the Fraction Collector III.
  • Functions as a preparative funnel adapter.
  • Compatible with the Fraction Collector III system.
  • Designed for multi-purpose laboratory applications.
  • Categorized as a spare part or kit.
Catalog No. 50-793-100 Supplier Waters Corp Supplier No. 725000113
The Multi-Purpose Rack is a versatile component designed for laboratory use, functioning as a preparative funnel adapter for the Fraction Collector III.
  • Functions as a preparative funnel adapter.
  • Compatible with the Fraction Collector III system.
  • Designed for multi-purpose laboratory applications.
  • Categorized as a spare part or kit.