Learn More
Alkali Scientific Multi Purpose Sample Cups for Clinical Analyzers, 1000/Pk, 4mL (17.26x37.90mm)
SDP
Catalog No. 5000662377
Encompass_Preferred
Polystyrene (PS) Multi Purpose Sample Cups are designed for use with clinical analyzers and automated testing instruments. Manufactured from high clarity polystyrene, these cups provide excellent optical transparency for accurate photometric and chemical analysis. Each cup is precision molded to ensure a secure fit in analyzer sample trays, supporting consistent sample positioning and reliable test results. Compatible with analyzers from Roche Cobas and Elecsys systems, Hitachi, Beckman, and other major platforms, these cups are suitable for handling serum, plasma, urine, calibrators, controls, and reagent samples. The smooth interior surface helps minimize sample retention, improving measurement consistency in clinical, research, and industrial laboratory workflows.
Catalog No. 50-006-62377 Supplier Alkali Scientific Supplier No. ASCMULTI04
Polystyrene (PS) Multi Purpose Sample Cups are designed for use with clinical analyzers and automated testing instruments. Manufactured from high clarity polystyrene, these cups provide excellent optical transparency for accurate photometric and chemical analysis. Each cup is precision molded to ensure a secure fit in analyzer sample trays, supporting consistent sample positioning and reliable test results. Compatible with analyzers from Roche Cobas and Elecsys systems, Hitachi, Beckman, and other major platforms, these cups are suitable for handling serum, plasma, urine, calibrators, controls, and reagent samples. The smooth interior surface helps minimize sample retention, improving measurement consistency in clinical, research, and industrial laboratory workflows.