Learn More
AccuTec Blades Multi-Purpose Scraping Tools
Catalog No. 17989136
Change view
Click to view available options
Product Type:
Heavy-duty Scraper
Catalog No. Product Type
17-989-136 Heavy-duty Scraper
1 options
This item is not returnable. View return policy
Catalog No. 17-989-136 Supplier AccuTec Blades Supplier No. 660445
This item is not returnable. View return policy
AccuTec Blades Multi-Purpose Scraping Tools available in two different sizes for a variety of applications
  • Replacable Blades
  • Contour Design
  • Brass Carrier with 3 Postions
  • Safe Scraping Stop
Specifications
Product Type Heavy-duty Scraper
Sterility Non-sterile
For Use With (Application) Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
For Use With (Equipment) Single-edge Blade
Includes 1 Replacement Blade
Length (English) 1.5 in.
Length (Metric) 38.10 mm
Material Carbon Steel
Quantity 1 Scraper
Thickness (Metric) 0.23 mm