Learn More
AccuTec Blades Multi-Purpose Scraping Tools
Catalog No. p-7182793
Change view
Click to view available options
Product Type:
Heavy-duty Scraper
Catalog No. Product Type
17-989-136 Heavy-duty Scraper
1 options
This item is not returnable. View return policy
Catalog No. 17-989-136 Supplier AccuTec Blades Supplier No. 660445
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
AccuTec Blades Multi-Purpose Scraping Tools available in two different sizes for a variety of applications
  • Replacable Blades
  • Contour Design
  • Brass Carrier with 3 Postions
  • Safe Scraping Stop
Specifications
Product Type Heavy-duty Scraper
Sterility Non-sterile
For Use With (Application) Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
For Use With (Equipment) Single-edge Blade
Includes 1 Replacement Blade
Length (English) 1.5 in.
Length (Metric) 38.10 mm
Material Carbon Steel
Quantity 1 Scraper
Thickness (Metric) 0.23 mm