Learn More
Research Products International Corp Multi-Purpose Vortexer
SDP
Catalog No. 502388446
Encompass
Multi-Purpose Vortexer is designed to hold a variety of tubes, containers, and microplates all within one mixing head design. Unit is capable of vortexing microplates and 6 different tube sizes with the convenience of a single mixing head attachment. Center pad area offers hand held mixing and performs standard tasks of a vortex mixer plus more. Features variable speed control (up to approx. 3000 rpm) and control switch with three operations (intermittent, off, continuous). Constructed of durable, easy to clean ABS with stable feet to dampen vibration.
One mixing head design
Variable speed control
Compact design
Five year warranty
Mixing Head Accommodates:
Center pad area for hand held mixing
Assay plate x 1 (to be used with retention cords)
50 mL conical tubes x 2 (horizontally mounted)
15 mL conical tubes x 2 (horizontally mounted)
Catalog No. 50-238-8446 Supplier Research Products International Corp Supplier No. 249250
Multi-Purpose Vortexer is designed to hold a variety of tubes, containers, and microplates all within one mixing head design. Unit is capable of vortexing microplates and 6 different tube sizes with the convenience of a single mixing head attachment. Center pad area offers hand held mixing and performs standard tasks of a vortex mixer plus more. Features variable speed control (up to approx. 3000 rpm) and control switch with three operations (intermittent, off, continuous). Constructed of durable, easy to clean ABS with stable feet to dampen vibration.
One mixing head design
Variable speed control
Compact design
Five year warranty
Mixing Head Accommodates:
Center pad area for hand held mixing
Assay plate x 1 (to be used with retention cords)
50 mL conical tubes x 2 (horizontally mounted)
15 mL conical tubes x 2 (horizontally mounted)