Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Multi-Sample Needles

Catalog No. p-7222362 Shop All Fisherbrand Products
Change view
Click to view available options
Gauge:
21 G
22 G
Preattached Holder:
No
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Preattached Holder
22-024-236 21 G No
22-024-235 22 G No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-024-236
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Fisherbrand Multi-sample Needle makes it possible to collect several samples with a single vein puncture.

  • Product Benefits:
    • Low-angle bevel design allows easy penetration
    • Color-coded caps and hubs allow easy identification of needle gauge size

Specifications

Product Type Multi-sample Needle
Sterility Sterile
Disposable Single-use
For Use With (Equipment) Fisherbrand Blood Collection Tube Holder
Gauge 21 G
Latex Free Yes
Length (English) 1 in.
Preattached Holder No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.