Learn More
BURKLE INC Multi sampler, Alu, 59.06 in. (150 cm), 5 chambers per 0.57 oz. (17 ml)
Catalog No. 5000026093
Encompass_Preferred
The multi-sampler with separate collection chambers is designed for simultaneous sampling from multiple zones or layers of bulk materials. Ideal for bags, drums, silos, big bags, or piles, it enables representative sampling from varying depths or areas, ensuring accurate material characterization. Individual target point samples can be analyzed separately to assess variability, or combined to create a composite sample for quality control. Suitable for vertical and horizontal use, the minimal gap between inner and outer tube reduces contamination risk, while the removable tip allows thorough cleaning to prevent cross-contamination. Color-coded caps help identify samplers for different materials. Built for durability, the robust construction and high-quality craftsmanship ensure long-lasting reliability and precise sampling for industrial, laboratory, and quality assurance applications.
Catalog No. 50-000-26093 Supplier BURKLE INC Supplier No. 53171150
The multi-sampler with separate collection chambers is designed for simultaneous sampling from multiple zones or layers of bulk materials. Ideal for bags, drums, silos, big bags, or piles, it enables representative sampling from varying depths or areas, ensuring accurate material characterization. Individual target point samples can be analyzed separately to assess variability, or combined to create a composite sample for quality control. Suitable for vertical and horizontal use, the minimal gap between inner and outer tube reduces contamination risk, while the removable tip allows thorough cleaning to prevent cross-contamination. Color-coded caps help identify samplers for different materials. Built for durability, the robust construction and high-quality craftsmanship ensure long-lasting reliability and precise sampling for industrial, laboratory, and quality assurance applications.