Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Multi-Specimen Collector Dispenser
SDP

Catalog No. 22000705
Change view
Click to view available options
Mounting:
Tape
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-705 Wall Mount
22-000-704 Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-705 Supplier Poltex Supplier No. SPECDISPW
Add to Cart
Add to Cart
This item is not returnable. View return policy

Multi-Specimen Collector Dispenser is used for holding commode specimen collectors and urine cups.

  • Holds commode specimen collectors and urine cups.
  • Durable, cleanable PETG construction.
  • See image for dimensions.
  • Choose from Wall Mount (keyhole slots on back), Powerful Magnets, or VHB tape mount.

Specifications

Autoclavable Not Autoclavable
Color Clear
Material PETG
Positions 5
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Urine Cups, Toilet Accessories
Mounting Wall Mount
Height (Metric) 325 in.
Width (Metric) 470 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.