Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Multi-Strain Human Papillomavirus (HPV) Verification Panel (Inactivated Pellet)
SDP

Catalog No. 23047369 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
20 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-369 20 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-369 Supplier Microbiologics™ Supplier No. 8235
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics offers a wide range of bundled QC Sets and Panels for the most commonly used technologies and standards. Each set or panel contains the manufacturer-recommended quality control strains for a particular instrument or test method in a simple, ready-to-use format.

Multi-Strain Human Papillomavirus (HPV) Verification Panel (Inactivated Pellet) contains 20 individually packaged inactivated, pre-pooled microorganism pellets comprised of human cells and the strains (20 vials total): Human Papillomavirus Type 16 surrogate 1, Human Papillomavirus Type 16 surrogate 2, Human Papillomavirus Type 18 surrogate 1, Human Papillomavirus Type 18 surrogate 2, Human Papillomavirus Type 31 surrogate 1, Human Papillomavirus Type 31 surrogate 2, Human Papillomavirus Type 45 surrogate 1, Human Papillomavirus Type 45 surrogate 2, Human Papillomavirus Type 68 surrogate 1, Human Papillomavirus Type 68 surrogate 2, and Human Cells

Specifications

Biosafety Level 1
Content And Storage 2°C to 8°C
Format QC Sets and Panels: Helix Elite™
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Microbial Identification
Packaging Type Plastic Box
Quantity 20 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.