Learn More
Electron Microscopy Sciences Core DX Multi Task Cart with Beige Drawers and Clear Door
SDP
Catalog No. 501927265
Encompass_Preferred
White Polyethylene Core DX Multi Tasking Storage Cart offers an efficient way to sort, store, organize, lock and transport a wide variety of supplies with drawers, shelves, built in glove & tissue dispensers and bulk storage features. Constructed from tough, non-reactive white high density polyethylene, ABS, and PETG this lab cart glides smoothly across most surfaces on its 4 heavy duty 3 inch non-marking rubber casters. The small footprint allows it to be stored securely and unobtrusively out of the way.
The cart features 3 full-extension drawers, 6 side shelves, 1 bulk storage area with locking door and two keys, and 2 with dispensing slots.
Color: Beige Drawers
Color: Clear Door
Catalog No. 50-192-7265 Supplier Electron Microscopy Sciences Supplier No. 6438627
White Polyethylene Core DX Multi Tasking Storage Cart offers an efficient way to sort, store, organize, lock and transport a wide variety of supplies with drawers, shelves, built in glove & tissue dispensers and bulk storage features. Constructed from tough, non-reactive white high density polyethylene, ABS, and PETG this lab cart glides smoothly across most surfaces on its 4 heavy duty 3 inch non-marking rubber casters. The small footprint allows it to be stored securely and unobtrusively out of the way.
The cart features 3 full-extension drawers, 6 side shelves, 1 bulk storage area with locking door and two keys, and 2 with dispensing slots.
Color: Beige Drawers
Color: Clear Door