Learn More
Research Products International Corp Multi-Therm Benchtop Incubator Shaker with Heating and Cooling
SDP
Catalog No. 501976836
Encompass
Multi-Therm combines all heating, cooling and shaking operations in one compact instrument that fits conveniently on a benchtop. Ideal for a variety of laboratory applications such as denaturing of nucleic acids and proteins; lipid extractions; bacteria and yeast cultivation; restriction digest; plasmid isolations and others. Easy to use and set with a sealed membrane keypad. Settings are continuously visible on a large LCD panel showing both actual and set values. Two year warranty. Heating Blocks available separately.
Blocks available separately for wide variety of tube sizes.
Catalog No. 50-197-6836 Supplier Research Products International Corp Supplier No. 247950
Multi-Therm combines all heating, cooling and shaking operations in one compact instrument that fits conveniently on a benchtop. Ideal for a variety of laboratory applications such as denaturing of nucleic acids and proteins; lipid extractions; bacteria and yeast cultivation; restriction digest; plasmid isolations and others. Easy to use and set with a sealed membrane keypad. Settings are continuously visible on a large LCD panel showing both actual and set values. Two year warranty. Heating Blocks available separately.
Blocks available separately for wide variety of tube sizes.