Learn More
Bellco Glass, Inc. Multi-Tier Complete, 0.5mL
SDP
Catalog No. 501945617
Encompass_Preferred
Description0.5mL MTP System ABS Plate with PTFE/Silicone/PTFE Liner, Cover & Glass 9x17mm Conical Vials. Product DetailsColor: NaturalVolume: 0.5mlGraduated: NoSilanized: NoLength Inches: 6Width Inches: 3.75Case Height Inches: 2.5Case Weight Lbs: 1Country Of Origin: USAHarmonization Code: 7017.20.0000Round Bottom: No
Catalog No. 50-194-5617 Supplier Bellco Glass, Inc. Supplier No. 451000100
Description0.5mL MTP System ABS Plate with PTFE/Silicone/PTFE Liner, Cover & Glass 9x17mm Conical Vials. Product DetailsColor: NaturalVolume: 0.5mlGraduated: NoSilanized: NoLength Inches: 6Width Inches: 3.75Case Height Inches: 2.5Case Weight Lbs: 1Country Of Origin: USAHarmonization Code: 7017.20.0000Round Bottom: No