Learn More
Bellco Glass, Inc. Multi-Tier Complete, 1.0mL
SDP
Catalog No. 501945618
Encompass_Preferred
Description1.0mL MTP System ABS Plate with PTFE/Silicone/PTFE Liner, Cover & Glass 9x30mm Conical VialsProduct DetailsColor: NaturalVolume: 1.0mlGraduated: NoSilanized: NoLength Inches: 6Width Inches: 3.75Case Height Inches: 2.5Case Weight Lbs: 1Country Of Origin: USAHarmonization Code: 7017.20.0000Round Bottom: No
Catalog No. 50-194-5618 Supplier Bellco Glass, Inc. Supplier No. 451000200
Description1.0mL MTP System ABS Plate with PTFE/Silicone/PTFE Liner, Cover & Glass 9x30mm Conical VialsProduct DetailsColor: NaturalVolume: 1.0mlGraduated: NoSilanized: NoLength Inches: 6Width Inches: 3.75Case Height Inches: 2.5Case Weight Lbs: 1Country Of Origin: USAHarmonization Code: 7017.20.0000Round Bottom: No