Learn More
Chemglass Life Sciences 1.5mL 96 Well Multi-Tier Reaction Plate, Topas, Complete
SDP
Catalog No. CG191503
Encompass_Preferred
1.5mL 96 Well Multi-Tier Reaction Plate, Topas, Complete. Our Multi-Tier 96 well reaction plates have a fixed height or can be multi-tiered to accept up to 1.5mL borosilicate glass inserts. Plates have the same footprint as standard 96 well polystyrene plates and are compatible with 96 well automation workstations. Complete setups include: Base Plate, Glass Inserts, Prescored PTFE/Silicone Septa, and Snap On Cover. PLEASE NOTE: PLATES ARE AUTOCLAVABLE.
Catalog No. CG191503 Supplier Chemglass Life Sciences Supplier No. CG191503
1.5mL 96 Well Multi-Tier Reaction Plate, Topas, Complete. Our Multi-Tier 96 well reaction plates have a fixed height or can be multi-tiered to accept up to 1.5mL borosilicate glass inserts. Plates have the same footprint as standard 96 well polystyrene plates and are compatible with 96 well automation workstations. Complete setups include: Base Plate, Glass Inserts, Prescored PTFE/Silicone Septa, and Snap On Cover. PLEASE NOTE: PLATES ARE AUTOCLAVABLE.