Learn More
Bellco Glass, Inc. Multi-Tier Universal Rack 35pl
SDP
Catalog No. 502330677
Encompass_Preferred
Universal Racks are multi-tier tube racks with four rows of holes arranged at four different levels. Specially designed for applications where space is limited but multiple size racks are required. Made of a heavy-duty high impact polymer, this rack is truly versatile.The highest row can accommodate five 30 or 50 mL centrifuge tubes in either the conical or freestanding configurationThe second row can hold eight 15 mL centrifuge tubesThe third row has ten holes for 12 × 75 mm tubesThe front row has 12 holes for microcentrifuge tubesDimensions: 8" (L) × 5 ⅖" (W) × 1 ¾" (H). Color-Yellow, Material-Polystyrene (PS), Package Type-Bulk, Carton Qty-4
Catalog No. 50-233-0677 Supplier Bellco Glass, Inc. Supplier No. 130000371
Add to Cart
Add to Cart
Universal Racks are multi-tier tube racks with four rows of holes arranged at four different levels. Specially designed for applications where space is limited but multiple size racks are required. Made of a heavy-duty high impact polymer, this rack is truly versatile.The highest row can accommodate five 30 or 50 mL centrifuge tubes in either the conical or freestanding configurationThe second row can hold eight 15 mL centrifuge tubesThe third row has ten holes for 12 × 75 mm tubesThe front row has 12 holes for microcentrifuge tubesDimensions: 8" (L) × 5 ⅖" (W) × 1 ¾" (H). Color-Yellow, Material-Polystyrene (PS), Package Type-Bulk, Carton Qty-4