Learn More
Associates of Cape Cod Inc MULTI-TST VIAL 0.06 EU/ML 25PK
SDP
Catalog No. NC0526497
Encompass
PYROTELL GEL CLOT LAL, 0.06 EU/mL SENSITIVITY, 5mL/VIAL, 25 VIALS/PACK
Catalog No. NC0526497 Supplier Associates of Cape Cod Inc Supplier No. G500625
PYROTELL GEL CLOT LAL, 0.06 EU/mL SENSITIVITY, 5mL/VIAL, 25 VIALS/PACK