Learn More
Research Products International Corp Multi-Tube Rack, 18 Holes
SDP
Catalog No. 501367681
Encompass
Multi-Tube Racks are ideal for holding centrifuge tubes including: (6) 50ml and (12) 15ml. Space saving design accommodates round and conical bottom tubes. Tube Racks are manufactured from polypropylene and are autoclavable.
Supplied flat and assembles quickly and easily.
Catalog No. 50-136-7681 Supplier Research Products International Corp Supplier No. 181056
Multi-Tube Racks are ideal for holding centrifuge tubes including: (6) 50ml and (12) 15ml. Space saving design accommodates round and conical bottom tubes. Tube Racks are manufactured from polypropylene and are autoclavable.
Supplied flat and assembles quickly and easily.