Learn More
Research Products International Corp Multi-Tube Rack, 35 Holes, Polypropylene, Autoclavable
SDP
Catalog No. 501367680
Encompass
Multi-Tube Racks are ideal for holding assorted size micro-tubes and centrifuge tubes including: 1.5, 2.0, 15 and 50 ml sizes. Space saving design accommodates round and conical bottom tubes. Tube Racks are manufactured from polypropylene and are autoclavable.
Supplied flat and assemble quickly and easily.
Catalog No. 50-136-7680 Supplier Research Products International Corp Supplier No. 181055
Multi-Tube Racks are ideal for holding assorted size micro-tubes and centrifuge tubes including: 1.5, 2.0, 15 and 50 ml sizes. Space saving design accommodates round and conical bottom tubes. Tube Racks are manufactured from polypropylene and are autoclavable.
Supplied flat and assemble quickly and easily.