Learn More
Chemglass Life Sciences Chemglass Adaptable Multi-Tube Rack, Blue/Green, Pack of 2 - for 12-30 mm (5-50 mL) tubes
SDP
Catalog No. NC2001552
Encompass
Pack of two adaptable multi-tube racks made from POM that securely hold a range of tube sizes. Each slot adapts to multiple diameters, the open-sided design permits label viewing, and racks can be linked to expand holding capacity; flat base keeps tubes upright and units are autoclavable.
  • Accommodates 12 mm-30 mm (5 mL-50 mL) tubes.
  • Flexible slots adapt to different tube diameters.
  • Holds tubes upright with a flat base.
  • Open sides allow label viewing.
  • Linkable design for increased capacity.
  • Made from POM and autoclavable.
  • Support under linked racks recommended when carrying.
Catalog No. NC2001552 Supplier Chemglass Life Sciences Supplier No. CLS4719001
Add to Cart
Add to Cart
Pack of two adaptable multi-tube racks made from POM that securely hold a range of tube sizes. Each slot adapts to multiple diameters, the open-sided design permits label viewing, and racks can be linked to expand holding capacity; flat base keeps tubes upright and units are autoclavable.
  • Accommodates 12 mm-30 mm (5 mL-50 mL) tubes.
  • Flexible slots adapt to different tube diameters.
  • Holds tubes upright with a flat base.
  • Open sides allow label viewing.
  • Linkable design for increased capacity.
  • Made from POM and autoclavable.
  • Support under linked racks recommended when carrying.