Description
- Offset connector allows the user to rotate the cartridge into 3 different positions to improves visibility and allow face shields or welding helmets to be worn without interference
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Specifications
| Material | Plastic |
| Product Type | Respirator Accessories |
| For Use With (Equipment) | Masks |
| Includes | Cartridges and filters |
| Quantity | 6/Box |