Learn More
Electron Microscopy Sciences Multi-Vial Multipurpose Containers
SDP
Catalog No. 50949215
Encompass_Preferred
Multi-vials are excellent multipurpose containers. They are primarily used for liquid scintillation counting, beta-gamma counting, and sample storage. They are made from a new type of polypropylene, which assures superior visibility. They come with white polyethylene caps and are perfect and convenient for easy marking. Features nominal volume of 5 ml, high density white polyethylene snap cap, and alternate white plug cap.
Catalog No. 50-949-215 Supplier Electron Microscopy Sciences Supplier No. 72659
Multi-vials are excellent multipurpose containers. They are primarily used for liquid scintillation counting, beta-gamma counting, and sample storage. They are made from a new type of polypropylene, which assures superior visibility. They come with white polyethylene caps and are perfect and convenient for easy marking. Features nominal volume of 5 ml, high density white polyethylene snap cap, and alternate white plug cap.