Learn More
Electron Microscopy Sciences Vortex-Genie Multi Vortex-Genie, 50 Hz, British Plug, 230 V
SDP
Catalog No. 501899720
Encompass_Preferred
A large capacity multi vessel vortex mixer that will vortex anything and everything. Same true vortexing as with the original Vortex-Genie 2. Holds multiple test tubes, vials, or cylinders of various sizes (racks included) or up to six 250 ml vessels, (more for smaller vessels) or a combination of tubes and vessels such as Erlenmeyer flasks. Designed and built "Genie™" strong, rugged and reliable. Digital control and display of speed and time for reproducibility and accuracy, with true speed display. Gentle ramp up to speed, so open containers will not spill. Use for gentle, orbital-style shaking or more vigorous vortexing. Durable and heavy zinc die casting with specially designed feet prevents "walking". No additional holders or accessories necessary. Racks are included. May be used in cold rooms or incubators.
Catalog No. 50-189-9720 Supplier Electron Microscopy Sciences Supplier No. 71846BR
A large capacity multi vessel vortex mixer that will vortex anything and everything. Same true vortexing as with the original Vortex-Genie 2. Holds multiple test tubes, vials, or cylinders of various sizes (racks included) or up to six 250 ml vessels, (more for smaller vessels) or a combination of tubes and vessels such as Erlenmeyer flasks. Designed and built "Genie™" strong, rugged and reliable. Digital control and display of speed and time for reproducibility and accuracy, with true speed display. Gentle ramp up to speed, so open containers will not spill. Use for gentle, orbital-style shaking or more vigorous vortexing. Durable and heavy zinc die casting with specially designed feet prevents "walking". No additional holders or accessories necessary. Racks are included. May be used in cold rooms or incubators.