Learn More
Electron Microscopy Sciences Transparent Wafer Container Multi, 2", Polypropylene Material
SDP
Catalog No. 501900226
Encompass_Preferred
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Containers have a storage capacity of 25 wafers and come with the base and the cover.
Use: 2" (50 mm)
Material: Polypropylene
Catalog No. 50-190-0226 Supplier Electron Microscopy Sciences Supplier No. 649182
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Containers have a storage capacity of 25 wafers and come with the base and the cover.
Use: 2" (50 mm)
Material: Polypropylene