Learn More
Velp™ MultiAluBlock™ Base
Description
- Used to perform numerous reactions reducing time and bench space
- For use with Velp Series 6 Hotplate Stirrers
Specifications
Specifications
| Description | Holds various combinations of MultiAluBlocks™ |
| Diameter (Metric) | 135 mm |
| For Use With (Equipment) | Series 6 Hotplate Stirrers |
| Type | Block Base |
| Warranty | 24 Months |
| Material | Aluminum |