Learn More
Alkali Scientific Western Blot MultiBoxes™ for Western Blotting, 4-Compartment Blot Box, each Compartment 32 x 58 x 28mm, Clear, 6/pk
SDP
Catalog No. 502389458
Encompass_Preferred
Western blot boxes and containers are available in clear. They fit nearly any sized blot and are available in common blot sizes and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Catalog No. 50-238-9458 Supplier Alkali Scientific Supplier No. BC1203
Western blot boxes and containers are available in clear. They fit nearly any sized blot and are available in common blot sizes and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.